Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthcare Diagnostics ADVIA Centaur TSH3-Ultra II

Catalog No. 21279174 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-279-174 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-279-174 Supplier Siemens Healthcare Diagnostics Supplier No. 11208702
Add to Cart
Add to Cart
This item is not returnable. View return policy

An assay for in vitro diagnostic use in the quantitative determination of thyroid-stimulating hormone (TSH, thyrotropin) in human serum and plasma.

An assay for in vitro diagnostic use in the quantitative determination of thyroid-stimulating hormone (TSH, thyrotropin) in human serum and plasma.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 5 ReadyPack primary reagent packs containing:
• TSH3ULII Lite Reagent, solid phase
• Ancillary Well Reagent
• CAL3A H/L, 2 vials

Store at 2°C to 8°C.
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes Thyroid Stimulating Hormone (TSH)
Detection Method Chemiluminescence
DoA Calibrators Yes
Sample Type Serum, EDTA plasma, Heparinized plasma
Type Assay
Disposable Single-use
Formulation BSA, BGG, goat serum, sodium azide (<0.1%), surfactant, preservatives
For Use With (Equipment) ADVIA Centaur XP/XPT
Linearity Assay is linear for the measuring interval of 0.010 to 150.000 μL U/mL (mL U/L). Linearity testing was performed in accordance with CLSI Document EP06-ed2.22.
Quantity 100 Tests
Analysis Time 18 Minutes
Form Liquid Ready-to-Use
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.