Learn More
Med Vet International Advantix Ii Blue, Dog, Over 55Lb, 6/Box, 6 Boxes
Catalog No. 502828548
Encompass_Preferred
Once-a-month topical solution for the prevention and treatment of ticks, fleas, mosquitoes, biting flies and lice on dogs. Active ingredients: imidacloprid 8.8%, permethrin 44.0%, pyriproxyfen 0.44% Repels and kills ticks including Deer ticks, American dog ticks, Brown dog ticks, and Lone Star ticks for up to 4 weeks Repels and kills mosquitoes for up to 4 weeks Kills fleas on dogs within 12 hours and continues to prevent infestations for 1 month Repels and kills all life stages of fleas Prevents development of fleas, flea eggs, pupae and larvae for 1 month Repels and prevents blood-feeding by biting flies Kills chewing lice Waterproof
Catalog No. 50-282-8548 Supplier Med Vet International Supplier No. PHADVTIXIIBL6
Once-a-month topical solution for the prevention and treatment of ticks, fleas, mosquitoes, biting flies and lice on dogs. Active ingredients: imidacloprid 8.8%, permethrin 44.0%, pyriproxyfen 0.44% Repels and kills ticks including Deer ticks, American dog ticks, Brown dog ticks, and Lone Star ticks for up to 4 weeks Repels and kills mosquitoes for up to 4 weeks Kills fleas on dogs within 12 hours and continues to prevent infestations for 1 month Repels and kills all life stages of fleas Prevents development of fleas, flea eggs, pupae and larvae for 1 month Repels and prevents blood-feeding by biting flies Kills chewing lice Waterproof