Learn More
Advnt Pro Strips™ 5 Professional Threat Detection Devices
SDP
Catalog No. 19166884
Encompass
Change view
Click to view available options
Product Type:
Threat Detection Test Strips
Catalog No. Product Type
19-166-884 Threat Detection Test Strips
1 options
Catalog No. 19-166-884 Supplier Advnt Supplier No. PS5T
Kit contains everything needed to properly perform test without any requirement of additional collection kits or expensive readers
  • Detects five separate threat agents: Anthrax, Ricin toxin, Botulinum toxin, Y. pestis (Plague) and SEB's
  • Includes all-in-one TripleThreat™ tube and instruction sheet
  • Each test packaged in protective, foil pouch
  • Sealed in red mylar pouch which doubles as bio-hazard bag
Specifications
Product Type Threat Detection Test Strips
For Use With (Application) Detecting Anthrax, Ricin, Botulinum, Y. Pestis, SEB
Quantity 10 Strips/Pouch