Learn More
Medchemexpress LLC HY-13058 5mg Medchemexpress, ADX-47273 CAS:851881-60-2 Purity:>98%
SDP
Catalog No. 502081244
Encompass_Preferred
Medchemexpress, HY-13058 5mg ADX-47273 CAS:851881-60-2 ADX-47273 is a potent, selective and brain-penetrant mGluR5 positive allosteric modulator (PAM), with an EC50 of 0.17 μM for potentiation of glutamate (50 nM) response. ADX-47273 has antipsychotic and procognitive activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1244 Supplier Medchemexpress LLC Supplier No. HY130585MG
Medchemexpress, HY-13058 5mg ADX-47273 CAS:851881-60-2 ADX-47273 is a potent, selective and brain-penetrant mGluR5 positive allosteric modulator (PAM), with an EC50 of 0.17 μM for potentiation of glutamate (50 nM) response. ADX-47273 has antipsychotic and procognitive activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.