Learn More
Cayman Chemical Fuziline, Purity: ≥95%, CAS: 80665-72-1, 10 mg
SDP
Catalog No. 502989016
Encompass_Preferred
A diterpene alkaloid with cardioprotective activity; reduces sodium pentobarbital-induced cell death in primary neonatal rat cardiomyocytes from 0.1-10 µM; reduces isoproterenol-induced production of ROS and apoptosis in H9c2 rat cardiomyocytes at 0.5 µM; reduces isoproterenol-induced myocardial necrosis and fibrosis in rats at 3 and 10 mg/kg
Catalog No. 50-298-9016 Supplier Cayman Chemical Supplier No. 3088910 MG
A diterpene alkaloid with cardioprotective activity; reduces sodium pentobarbital-induced cell death in primary neonatal rat cardiomyocytes from 0.1-10 µM; reduces isoproterenol-induced production of ROS and apoptosis in H9c2 rat cardiomyocytes at 0.5 µM; reduces isoproterenol-induced myocardial necrosis and fibrosis in rats at 3 and 10 mg/kg