Learn More
Medchemexpress LLC HY-19406 5mg Medchemexpress, AE-3763 CAS:291778-77-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY194065MG
Encompass_Preferred
Medchemexpress, HY-19406 5mg AE-3763 CAS:291778-77-3 AE-3763 is a peptide-based human neutrophil elastase inhibitor with an IC50 of 29 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2130