Learn More
Motic™ AE2000 Inverted LED Microscopes
Catalog No. 01258030 Shop All Motic Group Products
Change view
Click to view available options
Head Style:
Binocular
Trinocular
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style
01-258-030 Trinocular
01-258-029 Binocular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-258-030 Supplier Motic™ Supplier No. 1100103800267
Add to Cart
Add to Cart
This item is not returnable. View return policy
The AE2000 is a revolutionary design packed with features that set a new benchmark in affordable inverted microscopy. This inverted microscope is designed for routine live cell inspection with an easy and flexible set up, making it ideal for university, routine-clinical and lab environments.
  • 45° inclined Siedentopf head [Trinocular: (Light Split: 20/80)]
  • ELWD condenser with 0.3 numerical aperature
  • Phase slider Ph0, BF, Ph1
  • Centering telescope
  • Plain stage with metal and glass stage inserts
  • 3 W LED illumination with intensity control
  • Diopter Adjustment: ±5
Specifications
Magnification Power 10X
Objective Types CCIS™ PL Plan Achromat Objectives (PH4X, PH10X, PH20X)
Nosepiece Side-facing Quadruple
Head Style Trinocular
Lighting Type LED
Eyepiece WF 10X/20 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov