Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motic AE2000 Microscope and Camera Bundle

Catalog No. p-7228775 Shop All Motic Group Products
Change view
Click to view available options
Includes:
BMH4000X Camera Bundle
BTI10 Camera Bundle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-257-888 BMH4000X Camera Bundle
01-257-480 BTI10 Camera Bundle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-257-888 Supplier Motic Supplier No. 1100501700013
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The AE2000 is a revolutionary design that is packed with features and sets a new benchmark in affordable inverted microscopy. Motic’s AE2000 inverted microscope is designed for routine live cell inspection with an easy and flexible set up, ideal for university, routine-clinical, and lab environments.

  • 45° inclined Siedentopf head [Trinocular: (Light Split: 20/80)]
  • ELWD condenser with 0.3 numerical aperture
  • Phase slider Ph0, BF, Ph1
  • Centering telescope
  • Plain stage with metal and glass stage inserts
  • 3 W LED illumination with intensity control
  • Diopter Adjustment: ±5
  • PL PH 20x objective

Specifications

Includes BMH4000X Camera Bundle
Voltage 100/240 V
Base Style Plain stage with metal and glass stage inserts
Magnification Power 4X, 10X, 20X
Objective Types CCIS Plan Achromatic, CCIS Plan Achromat Phase
Nosepiece Left side facing quadruple
Weight (Metric) 12.2 kg
Dimensions (L x W x H) 556 x 218 x 496 mm
Condenser Type ELWD condenser N.A. 0.30 (WD = 72mm) with filter holder
Product Type Inverted Trinocular Microscope
Model AE2000 Trinocular Inverted Microscope LED
Head Style Siedentopf Trinocular, 360° Swiveling
Camera Type Moticam BMH4000X
Lighting Type Quartz Halogen, LED
Eyepiece N-WF 10X/20 mm, with diopter adjustment on both
Display Type LCD
Diaphragm Iris diaphragm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.