Learn More
Cayman Chemical Arachidonoyl Ethanolamide, 94421-68-8, 5 mg
SDP
Supplier:  Cayman Chemical 900505
Encompass_Preferred
An endogenous CB neurotransmitter that has Ki values ranging from 61 to 543 nM for CB1 receptors and from 279 to 1,940 nM for CB2 receptors
Catalog No. NC9419997