Learn More
Aviva Systems Biology Corp Aebp2 Antibody - C-terminal region (ARP39944_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39944_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Aebp2. It was validated on Western Blot. . Alternate names: AU023766, B230313N05RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-842