Learn More
Bioworld AEBSF, 500 mg
Catalog No. 50255358
Encompass_Preferred
AEBSF 4-(2-Aminoethyl)benzenesulfonyl Fluoride, HCl Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin and other related thrombolytic enzymes. Less toxic substitute for PMSF. Specific and irreversible inhibitor of serine proteases. Synonym: 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloridepKb: 9.19 (Predicted)Appearance: SolidMelting Point: 182-184 °C
Catalog No. 50-255-358 Supplier Bioworld Supplier No. 401600003
AEBSF 4-(2-Aminoethyl)benzenesulfonyl Fluoride, HCl Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin and other related thrombolytic enzymes. Less toxic substitute for PMSF. Specific and irreversible inhibitor of serine proteases. Synonym: 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloridepKb: 9.19 (Predicted)Appearance: SolidMelting Point: 182-184 °C