Learn More
Gold Biotechnology Inc AEBSF 10 g
SDP
Catalog No. 501532744
Encompass_Preferred
AEBSF is an irreversible serine protease inhibitor. AEBSF has been shown to inhibit trypsin, chymotrypsin, plasmin, kallikrein and thrombin. It inhibits by acylation of the active site of the enzyme. As an alternative to PMSF and DFP, AEBSF offers lower toxicity, improved solubility in water and improved stability in aqueous solutions. AEBSF has been used in cell culture in concentrations of up to 0.25mM. Aqueous solutions are stable between pH 5-6; limited stability above pH 7.5.
Catalog No. 50-153-2744 Supplier Gold Biotechnology Inc Supplier No. A54010
AEBSF is an irreversible serine protease inhibitor. AEBSF has been shown to inhibit trypsin, chymotrypsin, plasmin, kallikrein and thrombin. It inhibits by acylation of the active site of the enzyme. As an alternative to PMSF and DFP, AEBSF offers lower toxicity, improved solubility in water and improved stability in aqueous solutions. AEBSF has been used in cell culture in concentrations of up to 0.25mM. Aqueous solutions are stable between pH 5-6; limited stability above pH 7.5.