Learn More
Research Products International Corp AEBSF [4-(2-Aminoethyl)-benzenesulfonyl fluoride, HCl], 500 Milligrams
SDP
Catalog No. 50213115
Encompass_Preferred
Specific and irreversible inhibitor of serine proteases. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin and other related thrombolytic enzymes. Less toxic substitute for PMSF.
Catalog No. 50-213-115 Supplier Research Products International Corp Supplier No. A200100.5
Specific and irreversible inhibitor of serine proteases. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin and other related thrombolytic enzymes. Less toxic substitute for PMSF.