Learn More
Crescent Chemical Co Inc AEBSF-HCL RESEARCH GRADE
SDP
Catalog No. 50247284
Encompass_Preferred
AEBSF-HCl research grade Irreversible inhibitor of thrombin and other serine proteases (e.g. chymotrypsin, kallikrein, plasmin, proteinase K, trypsin). Inhibits by acylation of the active site of the enzyme. AEBSF is water-soluble and much less toxic than P
Catalog No. 50-247-284 Supplier Crescent Chemical Co Inc Supplier No. SE12745.03
AEBSF-HCl research grade Irreversible inhibitor of thrombin and other serine proteases (e.g. chymotrypsin, kallikrein, plasmin, proteinase K, trypsin). Inhibits by acylation of the active site of the enzyme. AEBSF is water-soluble and much less toxic than P