Learn More
Medchemexpress LLC Aebsf (hydrochloride) | 30827-99-7 | 99.87% | 10 MM 1 ML
SDP
Catalog No. 5000339464
Encompass_Preferred
AEBSF hydrochloride is an irreversible inhibitor of serine proteases such as chymotrypsin, kallikrein, plasmin, thrombin, and trypsin. It inhibits Aβ production by directly inhibiting β-secretase in various human cell lines and suppresses lysis of leukemic cells by human macrophages. It also interferes with blastocyst growth and HeLa cell adhesion. In vivo, AEBSF prolongs survival in *T. gondii* infection models and reduces inflammation in murine models.
  • Irreversible inhibitor of serine proteases (chymotrypsin, kallikrein, plasmin, thrombin, and trypsin).
  • Inhibits β-secretase and macrophage-mediated leukemic cell lysis.
  • Disturbs blastocyst growth and cell adhesion.
  • Supports research on *T. gondii* infection and inflammation.
  • Appearance: Solid, white to off-white.
Catalog No. 50-003-39464 Supplier Medchemexpress LLC Supplier No. HY1282110MM/1ML
AEBSF hydrochloride is an irreversible inhibitor of serine proteases such as chymotrypsin, kallikrein, plasmin, thrombin, and trypsin. It inhibits Aβ production by directly inhibiting β-secretase in various human cell lines and suppresses lysis of leukemic cells by human macrophages. It also interferes with blastocyst growth and HeLa cell adhesion. In vivo, AEBSF prolongs survival in *T. gondii* infection models and reduces inflammation in murine models.
  • Irreversible inhibitor of serine proteases (chymotrypsin, kallikrein, plasmin, thrombin, and trypsin).
  • Inhibits β-secretase and macrophage-mediated leukemic cell lysis.
  • Disturbs blastocyst growth and cell adhesion.
  • Supports research on *T. gondii* infection and inflammation.
  • Appearance: Solid, white to off-white.