Learn More
Medchemexpress LLC AEBSF hydrochloride | 30827-99-7 | 99.87% | 200 MG
SDP
Catalog No. 5000342882
Encompass_Preferred
AEBSF hydrochloride is an irreversible inhibitor of serine proteases, including chymotrypsin, kallikrein, plasmin, thrombin, and trypsin. It is for research use only and not sold to patients.
  • Inhibits the constitutive production of Aβ by directly inhibiting β-secretase in various human cell lines.
  • Inhibits the lysis of leukemic cells by human macrophages without affecting macrophage secretion of TNF-α and IL-1β.
  • Disturbs the growth of blastocysts on endometrial cells and inhibits the adhesion of HeLa cells on HUVECs by altering protein secretion.
  • Prolongs the survival of mice with lethal *T. gondii* infection *in vivo*.
  • Reduces airway response and inflammation in a cockroach allergen-induced murine model.
Catalog No. 50-003-42882 Supplier Medchemexpress LLC Supplier No. HY12821200MG
AEBSF hydrochloride is an irreversible inhibitor of serine proteases, including chymotrypsin, kallikrein, plasmin, thrombin, and trypsin. It is for research use only and not sold to patients.
  • Inhibits the constitutive production of Aβ by directly inhibiting β-secretase in various human cell lines.
  • Inhibits the lysis of leukemic cells by human macrophages without affecting macrophage secretion of TNF-α and IL-1β.
  • Disturbs the growth of blastocysts on endometrial cells and inhibits the adhesion of HeLa cells on HUVECs by altering protein secretion.
  • Prolongs the survival of mice with lethal *T. gondii* infection *in vivo*.
  • Reduces airway response and inflammation in a cockroach allergen-induced murine model.