Learn More
Cayman Chemical AEBSF (hydrochloride), Purity: ≥98%, CAS: 30827-99-7, 50 mg
SDP
Catalog No. 502955440
Encompass_Preferred
A water soluble, irreversible, broad spectrum inhibitor of serine proteases, including trypsin, chymotrypsin, plasmin, thrombin, and kallikreins; also inhibits NADPH oxidase
Catalog No. 50-295-5440 Supplier Cayman Chemical Supplier No. 1432150 MG
A water soluble, irreversible, broad spectrum inhibitor of serine proteases, including trypsin, chymotrypsin, plasmin, thrombin, and kallikreins; also inhibits NADPH oxidase