Learn More
Vector Laboratories AEC Peroxidase (HRP) Substrate Kit 300 ml
SDP
Catalog No. NC9821752
Encompass
AEC Peroxidase (HRP) Substrate Kit (3-amino-9-ethylcarbazole) produces a red reaction product that can be used as a single label or as a second color for multiple antigen labeling using either manual or automated IHC staining methods. Stained slides must be aqueously mounted (e.g. in VectaMount AQ). This kit contains stock solutions in convenient dropper bottles, including all of the reagents necessary to prepare about 300 ml of working solution (2-3000 sections). It can be also used for staining on blots. Vector Laboratories horseradish peroxidase (HRP) substrate kits include options that are cost-effective (e.g. standard DAB), sensitive (e.g. ImmPACT™ DAB) and convenient (ImmPACT DAB EqV). The range of vibrant colors available facilitates multiplex staining.
Catalog No. NC9821752 Supplier Vector Laboratories Supplier No. SK4200
AEC Peroxidase (HRP) Substrate Kit (3-amino-9-ethylcarbazole) produces a red reaction product that can be used as a single label or as a second color for multiple antigen labeling using either manual or automated IHC staining methods. Stained slides must be aqueously mounted (e.g. in VectaMount AQ). This kit contains stock solutions in convenient dropper bottles, including all of the reagents necessary to prepare about 300 ml of working solution (2-3000 sections). It can be also used for staining on blots. Vector Laboratories horseradish peroxidase (HRP) substrate kits include options that are cost-effective (e.g. standard DAB), sensitive (e.g. ImmPACT™ DAB) and convenient (ImmPACT DAB EqV). The range of vibrant colors available facilitates multiplex staining.