Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZOLL™ AED 3 Carry Case

Catalog No. 17300110 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
Carry Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-300-110 Carry Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-300-110 Supplier ZOLL™ Supplier No. 8000001250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gray and yellow ZOLL™ carry case

Holds:

  • ZOLL™ AED 3 with preconnected CPR Uni-Padz electrodes
  • CPR Uni-Padz

Compatible With:

  • ZOLL™ AED 3
  • ZOLL™ AED 3 BLS

Specifications

Product Type Carry Case
Color Gray, Yellow
For Use With (Equipment) ZOLL™ AED 3 with preconnected CPR Uni-Padz electrodes, CPR Uni-Padz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.