Learn More
ZOLL™ AED 3 Trainer
Catalog No. 17300112 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
AED 3 Trainer
Catalog No. Product Type
17-300-112 AED 3 Trainer
1 options
Catalog No. 17-300-112 Supplier ZOLL™ Supplier No. 802800000101
Add to Cart
Add to Cart
Demonstrates Real CPR Help with feedback, mimics clinical AED for students to experience
  • Active LCD display with full-color rescue images
  • Contact sensors in training pads detect pad contact with mannequin
  • On-device pause button allows instructor to pause scenario
  • Easy USB software update/language configuration
  • Real-time feedback for trainees
  • Teaches proper rate and depth of CPR chest compressions
Options:
  • Training scenario
  • Operating mode (semi or fully automatic)
  • Lay-rescuer prompts
  • Check-breathing prompts
  • Voice-prompt volume
Includes:
  • ZOLL™ AED 3 trainer device
  • ZOLL™ AED 3 trainer CPR Uni-Padz electrode training harness
  • Trainer electrode gels/liners for Uni-Padz
  • 4 D cell batteries
  • Operator's guide
Specifications
Product Type AED 3 Trainer
Includes ZOLL™ AED 3 trainer device, ZOLL™ AED 3 trainer CPR Uni-Padz electrode training harness, Trainer electrode gels/liner, Four D cell batteries, Operator's guide
For Use With (Application) For CPR training