Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZOLL™ AED Simulator

Catalog No. 17300116 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
ZOLL Simulator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-300-116 ZOLL Simulator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-300-116 Supplier ZOLL™ Supplier No. 8000000925
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creates shockable or non-shockable heart rhythms

For Use With:

  • ZOLL™ AED 3
  • ZOLL™ AED 3 BLS
  • AED Plus and/or AED Pro

Includes:

  • 9 V battery

Specifications

Product Type ZOLL Simulator
Color White
Includes 9 V battery
For Use With (Equipment) ZOLL™ AED 3, ZOLL™ AED 3 BLS, AED Plus and/or AED Pro
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.