Learn More
Selleck Chemical LLC AEE788 (NVP-AEE788) 5mg 497839-62-0
SDP
Supplier:  Selleck Chemical LLC S14865MG
Encompass_Preferred
AEE788 (NVP-AEE788) is a potent inhibitor of EGFR and HER2/ErbB2 with IC50 of 2 nM and 6 nM, less potent to VEGFR2/KDR, c-Abl, c-Src, and Flt-1, does not inhibit Ins-R, IGF-1R, PKC? and CDK1. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-677-4