Learn More
Medchemexpress LLC HY-W042156 500mg Medchemexpress, Aegeline CAS:456-12-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW042156500MG
Encompass_Preferred
Medchemexpress, HY-W042156 500mg Aegeline CAS:456-12-2 Aegeline, a main alkaloid, mimics the yeast SNARE protein Sec22p in suppressing α-synuclein and Bax toxicity in yeast. Aegeline restores growth of yeast cells suppressed by either αsyn or Bax. Antioxidant activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3478