Learn More
Medchemexpress LLC Aeide-C1-NHS ester | 824426-32-6 | 98.0% | C6H6N4O4 | 50 MG
SDP
Catalog No. 5000353520
Encompass_Preferred
Aeide-C1-NHS ester is an alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent, containing an Azide group, and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-53520 Supplier Medchemexpress LLC Supplier No. HY14075550MG
Aeide-C1-NHS ester is an alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent, containing an Azide group, and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups