Learn More
Medchemexpress LLC AEOL-10150 pentachloride | 286475-30-7 | 98.0% | 1033.24 | 50 MG
SDP
Catalog No. 5000385829
Encompass_Preferred
AEOL-10150 pentachloride is a metalloporphyrin-catalyzed antioxidant and a superoxide dismutase mimetic. It scavenges reactive oxygen species (ROS) and reactive nitrogen species (RNS), and modulates the NF-κB signaling pathway. This compound possesses potent antioxidant and anti-inflammatory activities, effectively mitigating tissue damage.
  • Scavenges ROS and RNS
  • Modulates NF-κB signaling pathway
  • Possesses potent antioxidant and anti-inflammatory activities
  • Mitigates tissue damage caused by radiation and chemical agents
  • Synergizes with radiotherapy to exert anticancer effects on prostate tumors
Catalog No. 50-003-85829 Supplier Medchemexpress LLC Supplier No. HY1941650MG
AEOL-10150 pentachloride is a metalloporphyrin-catalyzed antioxidant and a superoxide dismutase mimetic. It scavenges reactive oxygen species (ROS) and reactive nitrogen species (RNS), and modulates the NF-κB signaling pathway. This compound possesses potent antioxidant and anti-inflammatory activities, effectively mitigating tissue damage.
  • Scavenges ROS and RNS
  • Modulates NF-κB signaling pathway
  • Possesses potent antioxidant and anti-inflammatory activities
  • Mitigates tissue damage caused by radiation and chemical agents
  • Synergizes with radiotherapy to exert anticancer effects on prostate tumors