Learn More
Medchemexpress LLC Aeol-10150 pentachloride | 286475-30-7 | MFCD30532574 | 98.0% | 1033.24 g/mol | C48H56Cl5MnN12 | 10 MG
SDP
Catalog No. 5000297932
Encompass_Preferred
AEOL-10150 pentachloride is a metalloporphyrin catalytic antioxidant and superoxide dismutase (SOD) mimetic used in preclinical research. It scavenges reactive oxygen and nitrogen species, modulates NF-κB signaling, and has shown antioxidant, anti-inflammatory, and radioprotective effects in cellular and animal models.
  • Acts as a potent antioxidant and SOD mimetic.
  • Scavenges reactive oxygen species and reactive nitrogen species.
  • Modulates NF-κB signaling implicated in inflammation.
  • Demonstrated anti-inflammatory and radioprotective effects in preclinical models.
  • High purity (98.0%) suitable for research use.
  • Solid form, light brown to black, store sealed away from moisture.
  • Useful for studies of oxidative stress and radioprotection.
Catalog No. 50-002-97932 Supplier Medchemexpress LLC Supplier No. HY1941610MG
AEOL-10150 pentachloride is a metalloporphyrin catalytic antioxidant and superoxide dismutase (SOD) mimetic used in preclinical research. It scavenges reactive oxygen and nitrogen species, modulates NF-κB signaling, and has shown antioxidant, anti-inflammatory, and radioprotective effects in cellular and animal models.
  • Acts as a potent antioxidant and SOD mimetic.
  • Scavenges reactive oxygen species and reactive nitrogen species.
  • Modulates NF-κB signaling implicated in inflammation.
  • Demonstrated anti-inflammatory and radioprotective effects in preclinical models.
  • High purity (98.0%) suitable for research use.
  • Solid form, light brown to black, store sealed away from moisture.
  • Useful for studies of oxidative stress and radioprotection.