Learn More
TARGETMOL CHEMICALS INC AEOL-10150 pentachloride 5MG
SDP
Catalog No. 502879624
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AEOL-10150 pentachloride is a metalloporphyrin catalytic antioxidant and superoxide dismutase mimetic that protects lungs from radiation-induced injury.
Catalog No. 50-287-9624 Supplier TARGETMOL CHEMICALS INC Supplier No. T141335MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AEOL-10150 pentachloride is a metalloporphyrin catalytic antioxidant and superoxide dismutase mimetic that protects lungs from radiation-induced injury.