Learn More
Sino Biological Aequorea victoria GFP Tag PAb 20ul (Sino Biological)
Supplier:  Sino Biological 50714T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Aequorea victoria GFP (Catalog#13105-S07E; AAB65663; Ser2-Lys238). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-4515