Learn More
Research Products International Corp Aeraseal Sealing Film for Culture Plates, Sterile, 82 x 126 mm, 50 per Package
SDP
Catalog No. 50212463
Encompass
A 140um thick hydrophobic porous rayon sealing film including medical-grade adhesive for tissue culture plates, bio-blocks and 96-well plates where gas exchange is necessary for cell or bacterial growth. AeraSeal film minimizes cross-contamination between wells and reduces sample evaporation. Unique micro-porous design allows uniform air and CO2 exchange between wells for optimum cell growth or bacterial cultivation. This adhesive film provides a leak tight seal on culture plates or 96 well assay plates. Film can be easily pierced for sample recovery using a single or multi-channel pipettor.
Medical grade, non-cytotoxic adhesive has a thermal stability range of -20C to 80C. Removable tabs allow quick and easy positioning of film.
Sterile packaged 25 per sealed bag, two bags per case.
AeraSeal is a Trademark of Excel Scientific, Inc.
Removable, not reusable.
Catalog No. 50-212-463 Supplier Research Products International Corp Supplier No. 202505
A 140um thick hydrophobic porous rayon sealing film including medical-grade adhesive for tissue culture plates, bio-blocks and 96-well plates where gas exchange is necessary for cell or bacterial growth. AeraSeal film minimizes cross-contamination between wells and reduces sample evaporation. Unique micro-porous design allows uniform air and CO2 exchange between wells for optimum cell growth or bacterial cultivation. This adhesive film provides a leak tight seal on culture plates or 96 well assay plates. Film can be easily pierced for sample recovery using a single or multi-channel pipettor.
Medical grade, non-cytotoxic adhesive has a thermal stability range of -20C to 80C. Removable tabs allow quick and easy positioning of film.
Sterile packaged 25 per sealed bag, two bags per case.
AeraSeal is a Trademark of Excel Scientific, Inc.
Removable, not reusable.