Learn More
Thomas Scientific AeraSeal™ Film For Cell and Bacterial Culture Non Sterile
SDP
Catalog No. NC9740551
Encompass
Breathable Membranes for Cell and Bacteria CulturesPromotes uniform CO2/air exchange for each wellMinimizes outer well evaporation and cross contaminationEasily pierceable with pipet tips or pipets for sample recove
Catalog No. NC9740551 Supplier Thomas Scientific Supplier No. 6980A23
Breathable Membranes for Cell and Bacteria CulturesPromotes uniform CO2/air exchange for each wellMinimizes outer well evaporation and cross contaminationEasily pierceable with pipet tips or pipets for sample recove