Learn More
Andwin Scientific AERASEAL STERILE 50/PKAERASE
SDP
Catalog No. NC0122000
Encompass_Preferred
The AeraSeal sealing films are ideal for tissue culture plates, bio-blocks, and 96-well plates where the exchange of gas is needed for cell or bacterial growth. This product is 140 m thick and can be pierced with no difficulty using pipet tips or pipets for sample recovery. It can be used in temperatures ranging from -20C to 80C. The film is classified as non-cytotoxic and highly gas permeable. This product is sold in a pack of 50.
Catalog No. NC0122000 Supplier Andwin Scientific Supplier No. T242150
The AeraSeal sealing films are ideal for tissue culture plates, bio-blocks, and 96-well plates where the exchange of gas is needed for cell or bacterial growth. This product is 140 m thick and can be pierced with no difficulty using pipet tips or pipets for sample recovery. It can be used in temperatures ranging from -20C to 80C. The film is classified as non-cytotoxic and highly gas permeable. This product is sold in a pack of 50.