Learn More
Thomas Scientific AeraSeal Film, Sterile
SDP
Catalog No. NC0884352
Encompass
Breathable Membranes for Cell and Bacteria Cultures Promotes uniform CO2/air exchange for each well Minimizes outer well evaporation and cross contamination Easily pierceable with pipet tips or pipets for sample recovery Recommended for temperatures from -20°C to 80°C 140µm thick hydrophobic, breathable, sealing film with medical-grade adhesive for cell culture plates, bio-blocks, and 96-well plates. Non-cytotoxic. Film dimensions are 142.9 x 82.6 mm. Available non-sterile or sterile. Excel Scientific Sealing Film Selection Chart
Catalog No. NC0884352 Supplier Thomas Scientific Supplier No. 6980A25
Breathable Membranes for Cell and Bacteria Cultures Promotes uniform CO2/air exchange for each well Minimizes outer well evaporation and cross contamination Easily pierceable with pipet tips or pipets for sample recovery Recommended for temperatures from -20°C to 80°C 140µm thick hydrophobic, breathable, sealing film with medical-grade adhesive for cell culture plates, bio-blocks, and 96-well plates. Non-cytotoxic. Film dimensions are 142.9 x 82.6 mm. Available non-sterile or sterile. Excel Scientific Sealing Film Selection Chart