Learn More
Sigma Aldrich Fine Chemicals Biosciences AeraSealTM sterile
Catalog No. 502901003
Encompass
A 140 mm thick hydrophobic porous sealing film with medical-grade adhesive for tissue culture plates bio-blocks and 96-well plates where gas exchange is necessary for cell or bacterial growth. AeraSealA” sealing films minimize cross-contamination spillage and evaporation. They allow uniform air and CO2 exchange for all wells unlike plate lids which favor exchange for wells near the plate edges. Dimensions 82.6 by 142.9 mm for standard-size tissue culture platenon-cytotoxic highly gas permeable easily pierceable with pipet tips or pipets for sample recovery recommended for temperatures from -20 C to 80 CNote AeraSealA” films are suitable for hypoxic culture conditions
Catalog No. 50-290-1003 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z72157350EA
A 140 mm thick hydrophobic porous sealing film with medical-grade adhesive for tissue culture plates bio-blocks and 96-well plates where gas exchange is necessary for cell or bacterial growth. AeraSealA” sealing films minimize cross-contamination spillage and evaporation. They allow uniform air and CO2 exchange for all wells unlike plate lids which favor exchange for wells near the plate edges. Dimensions 82.6 by 142.9 mm for standard-size tissue culture platenon-cytotoxic highly gas permeable easily pierceable with pipet tips or pipets for sample recovery recommended for temperatures from -20 C to 80 CNote AeraSealA” films are suitable for hypoxic culture conditions