Learn More
Kuhner Shaker Inc AERATION CAP FOR USE WITH..125
Catalog No. 502570083
Encompass
Aeration cap for use with..125 mL, 250 mL, 500 mL and 1000 mL..Corning cell culture flasks;..not autoclavable
Catalog No. 50-257-0083 Supplier Kuhner Shaker Inc Supplier No. SMTO00006
Aeration cap for use with..125 mL, 250 mL, 500 mL and 1000 mL..Corning cell culture flasks;..not autoclavable