Learn More
Chemglass Life Sciences Chemglass Air Aeration Tube (M10) - 226 mm, for 1 L-2 L Bioreactors
SDP
Supplier:  Chemglass Life Sciences CLS137906
Encompass
Chemglass air aeration tube with M10 thread and 226 mm tube length, designed for 1 L and 2 L bioreactors. The drilled-hole sparger extends to the bottom of the vessel and terminates below the impellers to produce bubbles that are broken up by the impellers for improved oxygen transfer.
  • Designed for high gas flow rates in small bioreactors.
  • Drilled-hole sparger extends to vessel bottom for efficient gas delivery.
  • Terminates below impellers to improve bubble breakup and oxygen transfer.
  • 316L stainless steel with electropolished finish for corrosion resistance.
  • M10 threaded connection for secure mounting to headplates.
  • Compatible with 1 L and 2 L bioreactor vessels.
Catalog No. NC2821224