Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Bulk Packing, 1g

Catalog No. 502393157 Shop All Phenomenex Products
Encompass

Aeris™ 2.6 m PEPTIDE XB-C18 100 04A-4505

Catalog No. 50-239-3157 Supplier Phenomenex Inc Supplier No. 04A4505
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aeris™ 2.6 m PEPTIDE XB-C18 100 04A-4505

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.