Learn More
Malvern Panalytical AERO QAS STD 10 X 2.5G VIALS
Catalog No. NC3762691
Encompass
  • Convenient format of 10 vials
  • Each vial contains 2.5 g
  • Supports precise instrument calibration
  • Ensures high measurement accuracy
  • Reliable and consistent standard material
  • Crucial for maintaining data integrity
  • Enhances reproducibility in particle size analysis
  • Compatible with Mastersizer 3000
  • Lead time of 1-2 weeks
Catalog No. NC3762691 Supplier Malvern Panalytical Supplier No. CRM3500
  • Convenient format of 10 vials
  • Each vial contains 2.5 g
  • Supports precise instrument calibration
  • Ensures high measurement accuracy
  • Reliable and consistent standard material
  • Crucial for maintaining data integrity
  • Enhances reproducibility in particle size analysis
  • Compatible with Mastersizer 3000
  • Lead time of 1-2 weeks