Learn More
MSC Vikan Aero Tote Tub, Polyethylene, 82 gal, White, USDA, FDA Approved
Catalog No. 502837839
Encompass
This polyethylene tub is a well-constructed container designed for various food products. With an 82-gallon capacity, its versatile modular design allows for secure nesting, stacking, and in-plant moving with a forklift. Measuring 40.7 inches in length, 37 inches in width, and 20.7 inches in height, this tub can also be used with an optional lid and pallet.
  • High-density polyethylene construction for durability.
  • Versatile modular design for secure nesting, stacking, and forklift compatibility.
  • Suitable for protection, storage, and transportation of various food products including meat, seafood, poultry, and grains.
  • Meets all USDA and FDA specifications.
  • Designed for hygiene-critical environments in food manufacturing, retail, and service.
  • Easy to clean and maintain for optimal hygiene control.
  • Can be used with an optional lid and pallet.
Catalog No. 50-283-7839 Supplier MSC Supplier No. 53438792
This polyethylene tub is a well-constructed container designed for various food products. With an 82-gallon capacity, its versatile modular design allows for secure nesting, stacking, and in-plant moving with a forklift. Measuring 40.7 inches in length, 37 inches in width, and 20.7 inches in height, this tub can also be used with an optional lid and pallet.
  • High-density polyethylene construction for durability.
  • Versatile modular design for secure nesting, stacking, and forklift compatibility.
  • Suitable for protection, storage, and transportation of various food products including meat, seafood, poultry, and grains.
  • Meets all USDA and FDA specifications.
  • Designed for hygiene-critical environments in food manufacturing, retail, and service.
  • Easy to clean and maintain for optimal hygiene control.
  • Can be used with an optional lid and pallet.