Learn More
MSC Vikan Aero Tote Tub, Polyethylene with Drain Plug, 18 cu. ft.
Supplier:  MSC 47172580
Encompass
The Aero Tote Tub with Drain Plug is a well-constructed container made from high-density polyethylene, offering protection for various food products. Its versatile modular design allows for secure nesting, stacking, and in-plant moving with a forklift. This tub meets all USDA specifications and is commonly used in food manufacturing, food retail, restaurants, and food service. The integrated drain plug facilitates easy liquid management.
  • Durable high-density polyethylene construction
  • Hygienic one-piece design with no seams or crevices
  • Modular design allows for secure nesting, stacking, and forklift movement
  • Includes a drain plug for easy liquid management
  • Meets USDA specifications for food handling
  • Suitable for meat, seafood, poultry, and grain applications
  • Dishwasher safe
  • Autoclave safe
  • Made in America
Catalog No. 50-283-6261