Learn More
Vikan™ Aero Tote Tub
Catalog No. 17227671 Shop All Remco Products
Change view
Click to view available options
Color:
White
Catalog No. Color
17-227-671 White
1 options
Catalog No. 17-227-671 Supplier Vikan™ Supplier No. 6921
Add to Cart
Add to Cart
Aero Tote Tub is commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications.
  • High-density polyethylene walls offer protection for all kinds of food products
  • The versatile modular design allows for secure nesting, stacking and in-plant moving with a forklift
  • These tubs meet all USDA specifications and are commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications
  • The Aero-tote tub can be used with optional lid and pallet
Specifications
Product Type Aero Tub
Color White
Material Polyethylene
Certifications/Compliance FDA Compliant
Quantity 1
Dimensions (L x W x H) 40.7 x 37 x 20.7 in.