Learn More
ELITechGroup Aerospray Hematology Reagent D, Aerofix Fixative, 500mL
SDP
Catalog No. NC9544308
Encompass
This item has a minimum order quantity of 3 per supplier requirements. Hematology fixative solution; Liquid, Pale blue, characteristic alcohol odor; 500mL; Hematology Reagent D Aerofix Fixative; B.P. 65 deg.C; Freezing Point Approx. -100 deg.C; Storage: RT away from direct light
Catalog No. NC9544308 Supplier ELITechGroup Supplier No. SS048
This item has a minimum order quantity of 3 per supplier requirements. Hematology fixative solution; Liquid, Pale blue, characteristic alcohol odor; 500mL; Hematology Reagent D Aerofix Fixative; B.P. 65 deg.C; Freezing Point Approx. -100 deg.C; Storage: RT away from direct light