Learn More
ATCC Aeromonas hydrophila
Supplier:  ATCC 7966
Encompass
Aeromonas hydrophila strain [CDC 359-60, IAM 12460, NCIB 9240, NCMB 86, NCTC 8049, RH 250] was isolated from a tin of milk with a fishy odor. This bacterial type strain is whole-genome sequenced and can be used in water testing.
Catalog No. 50-238-3358