Learn More
ATCC Aeromonas hydrophila
Supplier:  ATCC 35654
Encompass
Aeromonas hydrophila strain LRA 3300 776 [API SA, DSM 6173] can be used as a quality control strain for API products.
Catalog No. 50-238-3276