Learn More
ATCC Bacillus thuringiensis
Catalog No. 502380993
Encompass
Bacillus thuringiensis strain USDA HD522 is a whole-genome sequenced strain that was isolated in Israel from sewage. This bacterial culture produces spores and toxic parasporal crystals. This bacterial culture produces Bacillus thuringiensis crystal protein delta-endotoxin.
Catalog No. 50-238-0993 Supplier ATCC Supplier No. 35646
Bacillus thuringiensis strain USDA HD522 is a whole-genome sequenced strain that was isolated in Israel from sewage. This bacterial culture produces spores and toxic parasporal crystals. This bacterial culture produces Bacillus thuringiensis crystal protein delta-endotoxin.