Learn More
Cayman Chemical -AeroplysInIn-1 100ug
SDP
Catalog No. 502986169
Encompass_Preferred
A sea sponge metabolite with diverse biological activities; induces cell death in MCF-7 and ZR-75-1 human breast cancer cells in a time- and concentration-dependent manner from 0.25-0.5 μM; blocks EGF-induced endocytosis of EGFR and EGF-dependent cell proliferation in MCF-7 cells; inhibits BAE cell proliferation and cord formation in a Matrigel™ assay; inhibits angiogenesis in a CAM assay from 1.5-3 nmol per egg; has antibacterial activity against Gram-positive bacteria; inhibits HIV-1 replication (IC50 = 14.6 μM)
Catalog No. 50-298-6169 Supplier Cayman Chemical Supplier No. 21498100 UG
A sea sponge metabolite with diverse biological activities; induces cell death in MCF-7 and ZR-75-1 human breast cancer cells in a time- and concentration-dependent manner from 0.25-0.5 μM; blocks EGF-induced endocytosis of EGFR and EGF-dependent cell proliferation in MCF-7 cells; inhibits BAE cell proliferation and cord formation in a Matrigel™ assay; inhibits angiogenesis in a CAM assay from 1.5-3 nmol per egg; has antibacterial activity against Gram-positive bacteria; inhibits HIV-1 replication (IC50 = 14.6 μM)