Learn More
Andwin Scientific AEROSEAL TM FILMS
SDP
Catalog No. NC9487614
Encompass_Preferred
The AeraSeal sealing films are ideal for tissue culture plates, bio-blocks, and 96-well plates where the exchange of gas is needed for cell or bacterial growth. This product is 140 m thick and can be pierced with no difficulty using pipet tips or pipets for sample recovery. It can be used in temperatures ranging from -20C to 80C. The film is classified as non-cytotoxic and highly gas permeable.
Catalog No. NC9487614 Supplier Andwin Scientific Supplier No. BS25
The AeraSeal sealing films are ideal for tissue culture plates, bio-blocks, and 96-well plates where the exchange of gas is needed for cell or bacterial growth. This product is 140 m thick and can be pierced with no difficulty using pipet tips or pipets for sample recovery. It can be used in temperatures ranging from -20C to 80C. The film is classified as non-cytotoxic and highly gas permeable.