Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Aerosol-Barrier Tips

Catalog No. 0270730 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 5 μL
0.2 to 10 μL
10 μL
100 μL
30 μL
50 μL
Format:
Bulk
Hinged Rack
Rack
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Format
02-707-30 0.2 to 10 μL Hinged Rack
02-707-152 0.1 to 5 μL Rack
02-707-33 30 μL Hinged Rack
02-707-161 100 μL Rack
02-707-27 0.2 to 10 μL Rack
02-707-153 10 μL Bulk
02-707-155 10 μL Hinged Rack
02-707-36 50 μL Hinged Rack
02-707-39 100 μL Hinged Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02-707-30

Prevent cross-contamination by using superior aerosol-barrier tips.

Fisherbrand™ Aerosol-Barrier tips can be used universally and protects the intergrity of your samples.

  • Unique small micron filter prevents aerosol contamination with no obstruction to airflow
  • Use aerosol barrier tips to ensure result integrity
  • Certified by independent laboratory RNase, DNase, DNA, and pyrogen free
  • Natural color

Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack, Sterile; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack, Sterile; SR: Stack Rack; IWS: Individually Wrapped, Sterile.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.2 to 10 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Length (English) 1.25 in.
Length (Metric) 3.17cm
Clean Claims DNase, RNase, DNA, and Pyrogen Free
Format Hinged Rack
Tip Style Barrier
For Use With (Equipment) Universal Fit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.