Learn More
MSC Justrite Aerosolv Standard System, Drum Mount, EPA Compliant, 1 ea
Catalog No. 502869651
Encompass
The Aerosolv system safely converts spent aerosol cans from solid hazardous waste to non-hazardous, recyclable steel. It accommodates industry-standard sized cans with a maximum height of 8.5 in (215 mm) and threads directly into a standard 2 in (51 mm) bung on 30- or 55-gallon drums. The body is constructed from low-maintenance aluminum, with all moving parts made of 308 stainless steel.
  • Converts spent aerosol cans from hazardous waste to recyclable steel
  • Mounts on 30- or 55-gallon drums
  • Includes two-piece coalescing filter and carbon cartridge
  • Angled pin ensures clean puncture
  • EPA compliant
  • Accepts cans up to 8.5 in (215 mm) height
  • Non-sparking puncture pin
  • Holds approximately 4,200 spent cans in a 55-gallon drum
  • Includes combination filter, safety goggles, and anti-static ground wire
Catalog No. 50-286-9651 Supplier MSC Supplier No. 51128064
The Aerosolv system safely converts spent aerosol cans from solid hazardous waste to non-hazardous, recyclable steel. It accommodates industry-standard sized cans with a maximum height of 8.5 in (215 mm) and threads directly into a standard 2 in (51 mm) bung on 30- or 55-gallon drums. The body is constructed from low-maintenance aluminum, with all moving parts made of 308 stainless steel.
  • Converts spent aerosol cans from hazardous waste to recyclable steel
  • Mounts on 30- or 55-gallon drums
  • Includes two-piece coalescing filter and carbon cartridge
  • Angled pin ensures clean puncture
  • EPA compliant
  • Accepts cans up to 8.5 in (215 mm) height
  • Non-sparking puncture pin
  • Holds approximately 4,200 spent cans in a 55-gallon drum
  • Includes combination filter, safety goggles, and anti-static ground wire