Learn More
Med Vet International Aerosol Inhalation Chamber Kit, J0843A, Each
Catalog No. 5000582126
Encompass_Preferred
• Used to deliver medication from metered dose inhaler • Also referred to as a "spacer", holds suspended medication until patient can readily inhale • Antistatic chamber is polypropylene with silicone connector • Soft silicone mask included Complete kit includes inhalation chamber and one mask of each size (S,M,L).
Catalog No. 50-005-82126 Supplier Med Vet International Supplier No. JOR843A
• Used to deliver medication from metered dose inhaler • Also referred to as a "spacer", holds suspended medication until patient can readily inhale • Antistatic chamber is polypropylene with silicone connector • Soft silicone mask included Complete kit includes inhalation chamber and one mask of each size (S,M,L).