Learn More
MSC LPS CFC Free Contact Cleaner, Plastic Safe, Flammable, 11 oz
Catalog No. 501112875
Encompass
LPS CFC Free Electro Contact Cleaner uses penetrating action to remove oil, grease, dirt, and flux from delicate precision mechanisms. It is designed to clean and degrease electronic components without leaving residue.
  • Safe on plastics
  • Fast evaporating, leaves no residue
  • NSF K2 certified for aerosol applications
  • Low odor
  • Flammable
  • RoHS compliant
  • Penetrating action removes oil, grease, dirt, and flux
  • Cleans and degreases delicate precision mechanisms
Catalog No. 50-111-2875 Supplier MSC Supplier No. 00252916
LPS CFC Free Electro Contact Cleaner uses penetrating action to remove oil, grease, dirt, and flux from delicate precision mechanisms. It is designed to clean and degrease electronic components without leaving residue.
  • Safe on plastics
  • Fast evaporating, leaves no residue
  • NSF K2 certified for aerosol applications
  • Low odor
  • Flammable
  • RoHS compliant
  • Penetrating action removes oil, grease, dirt, and flux
  • Cleans and degreases delicate precision mechanisms